3-Benzo[b]furylacetonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19550-03
L19550-MD
CAAAAL19550-03EA
225.94
CAD
CAAAAL19550-03
CAAAAL19550-MD
3-Benzo[b]furylacetonitrile 99%
3-Benzo[b]furylacetonitrile
Purity: 99%
CAS No.: 52407-43-9
Molecular Formula: C10H7NO
Formula Weight: 157.17
Melting Point: 38-39°
Boiling Point: 120-122°/4mm
Beilstein Reference: 1282785
MDL No.: MFCD02183555
Formula:
C₁₀H₇NO Boiling Pt: 120…122 °C/4 mm Melting Pt: 38…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD02183555 CAS Number: 52407-43-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...