2-Indanylboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(2,3-Dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
L19535-03
L19535-MD
CAAAAL19535-03EA
189.6
CAD
CAAAAL19535-03
CAAAAL19535-MD
2-Indanylboronic acid pinacol ester 95%
2-Indanylboronic acid pinacol ester
Purity: 95%
CAS No.: 608534-44-7
Molecular Formula: C15H21BO2
Formula Weight: 244.14
Refractive Index: 1.5040
MDL No.: MFCD03788726
Formula:
C₁₅H₂₁BO₂ MW: 244.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788726 CAS Number: 608534-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...