4-(Methylsulfonyl)phenylacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19504-06
L19504-14
CAAAAL19504-06EA
90.06
CAD
CAAAAL19504-06
CAAAAL19504-14
4-(Methylsulfonyl)phenylacetic acid 97%
4-(Methylsulfonyl)phenylacetic acid
Purity: 97%
CAS No.: 90536-66-6
Molecular Formula: C9H10O4S
Formula Weight: 214.24
Melting Point: 134-138°
Beilstein Reference: 2646649
MDL No.: MFCD00216495
Formula:
C₉H₁₀O₄S Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216495 CAS Number: 90536-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...