(R)-(-)-2-(Methoxymethyl)pyrrolidine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19492-03
L19492-06
L19492-MD
CAAAAL19492-03EA
216.46
CAD
CAAAAL19492-03
CAAAAL19492-06
CAAAAL19492-MD
(R)-(-)-2-(Methoxymethyl)pyrrolidine 98+%
(R)-(-)-2-(Methoxymethyl)pyrrolidine
Purity: 98+%
CAS No.: 84025-81-0
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 45°(113°F)
Refractive Index: 1.4457
Boiling Point: 61-62°/40mm
Beilstein Reference: 4229755
Optical Rotation: -2.2° (c=2 in benzene)
UN No.: 1993
MDL No.: MFCD00066219
Formula:
C₆H₁₃NO Boiling Pt: 61…62 °C/40 mm Density: 0.933 Flash Pt: 45 °C(113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066219 CAS Number: 84025-81-0 UN: 1993 ADR: "3",III |
Specification Test Results
Optical rotation | -2.2° (c=2 in benzene) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...