L-Proline-tert-butyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
L19489-03
L19489-06
CAAAAL19489-03EA
45.98
CAD
CAAAAL19489-03
CAAAAL19489-06
L-Proline-tert-butyl ester hydrochloride ≥98%
L-Proline-tert-butyl ester hydrochloride
Purity: 98+%
CAS No.: 5497-76-7
Molecular Formula: C9H17NO2.HCl
Formula Weight: 207.70
Melting Point: 110-112°
Beilstein Reference: 3596764
Optical Rotation: -31° (c=0.5 in water)
MDL No.: MFCD00153459
Formula:
C₉H₁₇NO₂ Melting Pt: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153459 CAS Number: 5497-76-7 EINECS: 226-835-2 |
Specification Test Results
Optical rotation | -31° (c=0.5 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...