Boc-Thz-OH 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-L-thiazolidine-4-carboxylic acid

L19488-03 L19488-06
CAAAAL19488-03EA 72.05 CAD
CAAAAL19488-03 CAAAAL19488-06
Boc-Thz-OH 98%
Boc-Thz-OH

Purity: 98% CAS No.: 51077-16-8 Molecular Formula: C9H15NO4S Formula Weight: 233.28 Melting Point: ca 133° dec. Beilstein Reference: 6686084 Optical Rotation: -113° (c=1 in methanol) MDL No.: MFCD00077002

Formula: C₉H₁₅NO₄S
MW: 233.282 g/mol
Melting Pt: ∼ 133 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00077002
CAS Number: 51077-16-8

Order Now

Specification Test Results

Optical rotation -113° (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR