N-Boc-1,6-hexanediamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-1-Boc-1,6-diaminohexane·HCl, Boc-1,6-diaminohexane·HCl, N-Boc-1,6-diaminohexane hydrochloride

L19487-03 L19487-06
CAAAAL19487-03EA 88.8 CAD
CAAAAL19487-03 CAAAAL19487-06
N-Boc-1,6-hexanediamine hydrochloride 98%
N-Boc-1,6-hexanediamine hydrochloride

Purity: 98% CAS No.: 65915-94-8 Molecular Formula: (CH3)3COCONH(CH2)6NH2.HCl Formula Weight: 252.79 Melting Point: ca 164° dec. Beilstein Reference: 3631740 MDL No.: MFCD00039072

Formula: C₁₁H₂₄N₂O₂·HCl
MW: 252.783 g/mol
Melting Pt: ∼ 164 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00039072
CAS Number: 65915-94-8
EINECS: 265-976-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR