N-Boc-ethylenediamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals

L19485-03 L19485-06
CAAAAL19485-03EA 170.64 CAD
CAAAAL19485-03 CAAAAL19485-06
N-Boc-ethylenediamine hydrochloride 98%
N-Boc-ethylenediamine hydrochloride

Purity: 98% CAS No.: 79513-35-2 Molecular Formula: (CH3)3COCONHCH2CH2NH2.HCl Formula Weight: 196.68 Melting Point: 148-151° Beilstein Reference: 3627511

Formula: C₇H₁₇ClN₂O₂
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD01076129
CAS Number: 79513-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR