Boc-Aib-OH ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: BOC-α-Methylalanine, N-Boc-alpha-methylalanine, 2-(Boc-amino)isobutyric Acid, Boc-Aib-OH

L19484-03 L19484-06
CAAAAL19484-03EA 41.08 CAD
CAAAAL19484-03 CAAAAL19484-06
Boc-Aib-OH ≥98%
Boc-Aib-OH

Purity: 98+% CAS No.: 30992-29-1 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Melting Point: 118-121° Beilstein Reference: 1953772 MDL No.: MFCD00042973

Formula: C₉H₁₇NO₄
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00042973
CAS Number: 30992-29-1
EINECS: 250-421-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR