2-(5-Ethylpyridin-2-yl)ethanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19482-06
L19482-14
CAAAAL19482-06EA
94.33
CAD
CAAAAL19482-06
CAAAAL19482-14
2-(5-Ethylpyridin-2-yl)ethanol 98%
2-(5-Ethylpyridin-2-yl)ethanol
Purity: 98%
CAS No.: 5223-06-3
Molecular Formula: C9H13NO
Formula Weight: 151.21
Melting Point: 39-44°
Beilstein Reference: 117340
MDL No.: MFCD00129040
Formula:
C₉H₁₃NO MW: 151.21 g/mol Melting Pt: 39…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00129040 CAS Number: 5223-06-3 EINECS: 226-024-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...