2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride

L19453-06 L19453-14
CAAAAL19453-06EA 62.09 CAD
CAAAAL19453-06 CAAAAL19453-14
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride 98%
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride

Purity: 98% CAS No.: 86604-75-3 Molecular Formula: C9H12ClNO.HCl Formula Weight: 222.12 Melting Point: ca 128° dec. Beilstein Reference: 5359162 MDL No.: MFCD00277472

Formula: C₉H₁₂ClNO
MW: 222.11 g/mol
Melting Pt: ca 128 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00277472
CAS Number: 86604-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR