Tetrachloropicolinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L19450-06 L19450-14
CAAAAL19450-06EA 69.52 CAD
CAAAAL19450-06 CAAAAL19450-14
Tetrachloropicolinic acid 98%
Tetrachloropicolinic acid

Purity: 98% CAS No.: 10469-09-7 Molecular Formula: C6HCl4NO2 Formula Weight: 260.89 Melting Point: 171-173° Beilstein Reference: 479080

Formula: C₆HCl₄NO₂
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00185833
CAS Number: 10469-09-7
EINECS: 233-956-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR