N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19412-03
L19412-06
CAAAAL19412-03EA
67.31
CAD
CAAAAL19412-03
CAAAAL19412-06
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 94%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Purity: 94%
CAS No.: 93102-05-7
Molecular Formula: C13H23NOSi
Formula Weight: 237.42
Storage Temperature: 2°C to 8°C
Flash Point: 66°(150°F)
Refractive Index: 1.4920
Boiling Point: 76°/0.3mm
Beilstein Reference: 4311216
MDL No.: MFCD00674005
Formula:
C₁₃H₂₃NOSi Boiling Pt: 76 °C/0.3 mm Density: 0.928 Flash Pt: 66 °C(150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00674005 CAS Number: 93102-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...