(S)-(+)-2-Amino-1-phenylethanol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19404-03
L19404-06
CAAAAL19404-03EA
134.77
CAD
CAAAAL19404-03
CAAAAL19404-06
(S)-(+)-2-Amino-1-phenylethanol 97%
(S)-(+)-2-Amino-1-phenylethanol
Purity: 97%
CAS No.: 56613-81-1
Molecular Formula: C8H11NO
Formula Weight: 137.18
Melting Point: 57-63°
Boiling Point: 160°/17mm
Beilstein Reference: 2412760
Optical Rotation: +42° (c=2 in ethanol)
UN No.: 3259
MDL No.: MFCD00239405
Formula:
C₈H₁₁NO Boiling Pt: 160 °C/17 mm Melting Pt: 57…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239405 CAS Number: 56613-81-1 UN: 3259 ADR: "8",III |
Specification Test Results
Optical rotation | +42° (c=2 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...