(R)-2-Amino-1-phenylethanol 97%, ee 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(-)-2-Amino-1-phenylethanol, (R)-(-)-alpha-(Aminomethyl)benzyl alcohol
L19403-03
L19403-06
CAAAAL19403-03EA
115.97
CAD
CAAAAL19403-03
CAAAAL19403-06
(R)-2-Amino-1-phenylethanol 97%, ee 98%
(R)-2-Amino-1-phenylethanol
Purity: 97%
CAS No.: 2549-14-6
Molecular Formula: C8H11NO
Formula Weight: 137.18
Odor: Amine-like
Melting Point: 57-63°
Boiling Point: 160°/17mm
Beilstein Reference: 3196196
Optical Rotation: -43° (c=2 in ethanol)
UN No.: 3259
MDL No.: MFCD00239406
Formula:
C₈H₁₁NO Boiling Pt: 160 °C/17 mm Melting Pt: 57…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239406 CAS Number: 2549-14-6 UN: 3259 ADR: "8",III |
Specification Test Results
Optical rotation | -43° (c=2 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...