Bis(pyridine)iodonium tetrafluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19385-03
L19385-MD
CAAAAL19385-03EA
101.59
CAD
CAAAAL19385-03
CAAAAL19385-MD
Bis(pyridine)iodonium tetrafluoroborate 97%
Bis(pyridine)iodonium tetrafluoroborate
Purity: 97%
CAS No.: 15656-28-7
Molecular Formula: C10H10BF4IN2
Formula Weight: 371.91
Storage Temperature: 2°C to 8°C
Odor: Odorless
Beilstein Reference: 4284739
MDL No.: MFCD03703393
Formula:
C₁₀H₁₀BF₄IN₂ MW: 371.91 g/mol Melting Pt: ca 160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03703393 CAS Number: 15656-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...