Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19384-03
L19384-06
CAAAAL19384-03EA
70.31
CAD
CAAAAL19384-03
CAAAAL19384-06
Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Purity: 97%
CAS No.: 132705-51-2
Molecular Formula: C12H24BrF6N3P2
Formula Weight: 466.19
Storage Temperature: -30°C to -10°C
Formula:
C₁₂H₂₄BrF₆N₃P₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00077412 CAS Number: 132705-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...