4-Nitrophenylketoxime on polystrene 0.8 - 1.0 mmol/g, cross-linked with 2% DVB 200-400 mesh
Supplier: Thermo Scientific Chemicals
L19376-03 L19376-06
CAAAAL19376-03EA 73.47 CAD
CAAAAL19376-03 CAAAAL19376-06
4-Nitrophenylketoxime on polystrene 0.8 - 1.0 mmol/g, cross-linked with 2% DVB 200-400 mesh
4-Nitrophenylketoxime on polystrene
MDL No.: MFCD00165078