3-Acetyl-5-bromopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L19362-03 L19362-06
CAAAAL19362-03EA 62.41 CAD
CAAAAL19362-03 CAAAAL19362-06
3-Acetyl-5-bromopyridine 97%
3-Acetyl-5-bromopyridine

Purity: 97% CAS No.: 38940-62-4 Molecular Formula: C7H6BrNO Formula Weight: 200.03 Melting Point: 77-79° Beilstein Reference: 112808

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD03086033
CAS Number: 38940-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR