(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Boc-Homopro-OH, 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, Boc-HomoPro-OH, 1-Boc-L-pipecolic Acid, (S)-1-Boc-2-piperidinecarboxylic Acid, Boc-Pip-OH, (S)-N-BOC-Piperidine-2-carboxylic acid
L19361-03
L19361-06
CAAAAL19361-03EA
156.1
CAD
CAAAAL19361-03
CAAAAL19361-06
CAAAAL19361-MD
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Purity: 98+%
CAS No.: 26250-84-0
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Melting Point: 121-125°
Beilstein Reference: 3652038
Optical Rotation: -46° (c=1 in methanol)
MDL No.: MFCD00151904
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol Melting Pt: 121…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151904 CAS Number: 26250-84-0 |
Specification Test Results
Optical rotation | -46° (c=1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...