1-Formyl-4-methylpiperazine 98%

Supplier: Thermo Scientific Chemicals
Warning

L19358-06 L19358-14
CAAAAL19358-06EA 65.73 CAD
CAAAAL19358-06 CAAAAL19358-14
1-Formyl-4-methylpiperazine 98%
1-Formyl-4-methylpiperazine

Purity: 98% CAS No.: 7556-55-0 Molecular Formula: C6H12N2O Formula Weight: 128.17 Flash Point: 110°(230°F) Refractive Index: 1.4910 Boiling Point: 236-238° Beilstein Reference: 113020 MDL No.: MFCD00085930

Formula: C₆H₁₂N₂O
Boiling Pt: 236…238 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00085930
CAS Number: 7556-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR