N-Boc-L-leucinol 97%

Supplier: Thermo Scientific Chemicals

L19352-03 L19352-06
CAAAAL19352-03EA 97.49 CAD
CAAAAL19352-03 CAAAAL19352-06
N-Boc-L-leucinol 97%
N-Boc-L-leucinol

Purity: 97% CAS No.: 82010-31-9 Molecular Formula: C11H23NO3 Formula Weight: 217.31 Flash Point: 110°(230°F) Refractive Index: 1.4500 Boiling Point: 212-214° Beilstein Reference: 4178460 Optical Rotation: -27.5° (c=2 in methanol) MDL No.: MFCD00076950

Formula: C₁₁H₂₃NO₃
Boiling Pt: 212…214 °C
Density: 0.980
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00076950
CAS Number: 82010-31-9

Order Now

Specification Test Results

Optical rotation -27.5° (c=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR