4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxy-3-pyrrolin-2-one, 4-Methoxy-1H-pyrrol-2(5H)-one

L19344-03 L19344-06
CAAAAL19344-03EA 118.82 CAD
CAAAAL19344-03 CAAAAL19344-06
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 99%
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one

Purity: 99% CAS No.: 69778-83-2 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Melting Point: 130-134° Beilstein Reference: 1524505 MDL No.: MFCD00071564

Formula: C₅H₇NO₂
MW: 113.12 g/mol
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00071564
CAS Number: 69778-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR