3-Fluorosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

L19324-03 L19324-06
CAAAAL19324-03EA 213.3 CAD
CAAAAL19324-03 CAAAAL19324-06 CAAAAL19324-MD
3-Fluorosalicylaldehyde 98%
3-Fluorosalicylaldehyde

Purity: 98% CAS No.: 394-50-3 Molecular Formula: C7H5FO2 Formula Weight: 140.12 Melting Point: 67-71° Beilstein Reference: 1932806

Formula: C₇H₅FO₂
MW: 140.12 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00003319
CAS Number: 394-50-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR