(S)-(+)-2-Aminononane ≥99%, ee 99%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

L19311-03 L19311-06 L19311-14
CAAAAL19311-03EA 82 CAD
CAAAAL19311-03 CAAAAL19311-06 CAAAAL19311-14
(S)-(+)-2-Aminononane ≥99%, ee 99%, ChiPros®
(S)-(+)-2-Aminononane

Purity: 99+% CAS No.: 13205-58-8 Molecular Formula: C9H21N Formula Weight: 143.27 Flash Point: 71°(159°F) Refractive Index: 1.4271 Boiling Point: 73°/19mm Optical Rotation: +6° (neat) UN No.: 2735 MDL No.: MFCD03844738

Formula: C₉H₂₁N
Boiling Pt: 73 °C/19 mm
Density: 0.782
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD03844738
CAS Number: 13205-58-8
EINECS: 663-044-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Optical rotation +6° (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR