4-Ethynyl-benzoic acid sodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 4-ethynylbenzoate

L19292-03 L19292-06
CAAAAL19292-03EA 147.89 CAD
CAAAAL19292-03 CAAAAL19292-06 CAAAAL19292-MD
4-Ethynyl-benzoic acid sodium salt 97%
4-Ethynyl-benzoic acid sodium salt

Purity: 97% CAS No.: 144693-65-2 Molecular Formula: C9H5NaO2 Formula Weight: 168.13 Beilstein Reference: 5836538 MDL No.: MFCD03426026

Formula: C₉H₅NaO₂
MW: 168.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426026
CAS Number: 144693-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR