L-Proline methyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Pro-OMe.HCl, H-Pro-OMe·HCl
L19290-03
L19290-06
CAAAAL19290-03EA
46.14
CAD
CAAAAL19290-03
CAAAAL19290-06
L-Proline methyl ester hydrochloride ≥98%
L-Proline methyl ester hydrochloride
Purity: 98+%
CAS No.: 2133-40-6
Molecular Formula: C6H11NO2.HCl
Formula Weight: 165.62
Storage Temperature: 2°C to 8°C
Melting Point: 71-78°
Beilstein Reference: 3596045
Optical Rotation: -33° (c=1 in water)
MDL No.: MFCD00012708
Formula:
C₆H₁₁NO₂·HCl Melting Pt: 71…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012708 CAS Number: 2133-40-6 EINECS: 218-363-0 |
Specification Test Results
Optical rotation | -33° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...