5-(3-Methylphenyl)-1H-tetrazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19263-03
L19263-MD
CAAAAL19263-03EA
152.79
CAD
CAAAAL19263-03
CAAAAL19263-MD
5-(3-Methylphenyl)-1H-tetrazole 99%
5-(3-Methylphenyl)-1H-tetrazole
Purity: 99%
CAS No.: 3441-00-7
Molecular Formula: C8H8N4
Formula Weight: 160.18
Melting Point: 143-146°
Beilstein Reference: 131205
Formula:
C₈H₈N₄ Melting Pt: 143…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093096 CAS Number: 3441-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...