5-(2-Methylphenyl)-1H-tetrazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19262-03
L19262-MD
CAAAAL19262-03EA
141.57
CAD
CAAAAL19262-03
CAAAAL19262-MD
5-(2-Methylphenyl)-1H-tetrazole 99%
5-(2-Methylphenyl)-1H-tetrazole
Purity: 99%
CAS No.: 51449-86-6
Molecular Formula: C8H8N4
Formula Weight: 160.18
Melting Point: 151-154°
Beilstein Reference: 136151
Formula:
C₈H₈N₄ Melting Pt: 151…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093095 CAS Number: 51449-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...