3-(Benzyloxy)phenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19242-03
L19242-MD
CAAAAL19242-03EA
189.6
CAD
CAAAAL19242-03
CAAAAL19242-MD
3-(Benzyloxy)phenyl isothiocyanate 97%
3-(Benzyloxy)phenyl isothiocyanate
Purity: 97%
CAS No.: 206559-36-6
Molecular Formula: C14H11NOS
Formula Weight: 241.31
Melting Point: 49-51°
UN No.: 2923
MDL No.: MFCD00060374
Formula:
C₁₄H₁₁NOS Melting Pt: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060374 CAS Number: 206559-36-6 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...