Tetrathiafulvalene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2'-Bi(1,3-dithiolylidene), TTF

L19229-03 L19229-MD
CAAAAL19229-03EA 279.66 CAD
CAAAAL19229-03 CAAAAL19229-MD
Tetrathiafulvalene 97%
Tetrathiafulvalene

Purity: 97% CAS No.: 31366-25-3 Molecular Formula: C6H4S4 Formula Weight: 204.35 Melting Point: 116-120° Beilstein Reference: 1617956 Merck Reference: 14,9242 MDL No.: MFCD00005492

Formula: C₆H₄S₄
MW: 204.356 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00005492
CAS Number: 31366-25-3
EINECS: 250-593-7
Merck Index: 14,9242

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR