2-Thiopheneacetamide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19221-06
L19221-14
L19221-22
CAAAAL19221-06EA
42.5
CAD
CAAAAL19221-06
CAAAAL19221-14
CAAAAL19221-22
2-Thiopheneacetamide 98%
2-Thiopheneacetamide
Purity: 98%
CAS No.: 4461-29-4
Molecular Formula: C6H7NOS
Formula Weight: 141.19
Melting Point: 145-148°
Beilstein Reference: 114558
MDL No.: MFCD00017556
Formula:
C₆H₇NOS Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017556 CAS Number: 4461-29-4 EINECS: 224-714-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...