2-(4-Benzyloxyphenyl)ethanol ≥98%
Supplier: Thermo Scientific Chemicals
L19217-06
L19217-14
CAAAAL19217-06EA
76
CAD
CAAAAL19217-06
CAAAAL19217-14
2-(4-Benzyloxyphenyl)ethanol ≥98%
2-(4-Benzyloxyphenyl)ethanol
Purity: 98+%
CAS No.: 61439-59-6
Molecular Formula: C15H16O2
Formula Weight: 228.29
Melting Point: 85-87°
Beilstein Reference: 2054624
MDL No.: MFCD00017532
Formula:
C₁₅H₁₆O₂ Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017532 CAS Number: 61439-59-6 EINECS: 262-795-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...