(1R)-(-)-Menthyl glyoxylate monohydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

L19208-14 L19208-22
CAAAAL19208-06EA 107.19 CAD
CAAAAL19208-06 CAAAAL19208-14 CAAAAL19208-22
(1R)-(-)-Menthyl glyoxylate monohydrate 98%
(1R)-(-)-Menthyl glyoxylate monohydrate

Purity: 98% CAS No.: 111969-64-3 Molecular Formula: C12H22O4 Formula Weight: 230.31 (212.29anhy) Melting Point: 76-80° Boiling Point: 95-96°/0.3mm Beilstein Reference: 5637371 Optical Rotation: -75° (c=1 in methanol) UN No.: 3077 MDL No.: MFCD00792503

Formula: C₁₂H₂₂O₄
MW: 230.31 g/mol (212.29 anhy)
Boiling Pt: 95…96 °C/0.3 mm
Melting Pt: 76…80 °C
Density: 1.182
Storage Temperature: Ambient
MDL Number: MFCD00792503
CAS Number: 111969-64-3
EINECS: 416-810-6
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR