Ethyl glyoxylate ∼50% in toluene
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19207-22
L19207-36
CAAAAL19207-22EA
111.39
CAD
CAAAAL19207-22
CAAAAL19207-36
Ethyl glyoxylate ∼50% in toluene
Ethyl glyoxylate
CAS No.: 924-44-7
Molecular Formula: OHCCO2CH2CH3
Formula Weight: 102.09
Flash Point: 7°(44°F)
Refractive Index: 1.4750
Boiling Point: 110°
UN No.: 1993
MDL No.: MFCD00044009
Formula:
C₄H₆O₃ Boiling Pt: 110 °C Density: 1.003 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044009 CAS Number: 924-44-7 EINECS: 213-105-3 UN: 1294 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...