Methyl-3-quinuclidinecarboxylate hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L19132-03 L19132-MD
CAAAAL19132-03EA 314.42 CAD
CAAAAL19132-03 CAAAAL19132-MD
Methyl-3-quinuclidinecarboxylate hydrochloride ≥98%
Methyl-3-quinuclidinecarboxylate hydrochloride

Purity: 98+% CAS No.: 54954-73-3 Molecular Formula: C9H15NO2.HCl Formula Weight: 205.69 Melting Point: ca 160° dec. Beilstein Reference: 4272456 MDL No.: MFCD01073582

Formula: C₉H₁₅NO₂·HCl
Melting Pt: ∼ 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01073582
CAS Number: 54954-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR