2-Methylimidazo[4,5-c]pyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-3H-imidazo[4,5-c]pyridine

L19131-03 L19131-06 L19131-MD
CAAAAL19131-03EA 105.39 CAD
CAAAAL19131-03 CAAAAL19131-06 CAAAAL19131-MD
2-Methylimidazo[4,5-c]pyridine 99%
2-Methylimidazo[4,5-c]pyridine

Purity: 99% CAS No.: 63604-59-1 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 176-178° Beilstein Reference: 3234

Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD03426025
CAS Number: 63604-59-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR