Dichloro(p-cymene)ruthenium(II) dimer 98%

Supplier: Thermo Scientific Chemicals
Danger

L19126-04 L19126-09 L19126-ME
CAAAAL19126-04EA 164.32 CAD
CAAAAL19126-04 CAAAAL19126-09 CAAAAL19126-ME
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer

Fieser: 19,120 21,52


Purity: 98% CAS No.: 52462-29-0 Molecular Formula: C20H28Cl4Ru2 Formula Weight: 612.40 MDL No.: MFCD00064793

Formula: C₂₀H₂₈Cl₄Ru₂
Storage Temperature: Ambient
MDL Number: MFCD00064793
CAS Number: 52462-29-0
EINECS: 435-530-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR