5-Methoxy-1H-indole-3-carbaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

L19072-03 L19072-06
CAAAAL19072-03EA 140.62 CAD
CAAAAL19072-03 CAAAAL19072-06 CAAAAL19072-MD
5-Methoxy-1H-indole-3-carbaldehyde 99%
5-Methoxy-1H-indole-3-carbaldehyde

Purity: 99% CAS No.: 10601-19-1 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 179-183° Beilstein Reference: 132769 MDL No.: MFCD00005623

Formula: C10H9NO2
MW: 175.19 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00005623
CAS Number: 10601-19-1
EINECS: 234-220-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR