(R)-(-)-α-Cyclohexanemethylamine 98%, ee 94%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(-)-1-Cyclohexylethylamine, (R)-1-Cyclohexylethylamine
L19051-03
L19051-06
CAAAAL19051-03EA
54.98
CAD
CAAAAL19051-03
CAAAAL19051-06
CAAAAL19051-14
(R)-(-)-α-Cyclohexanemethylamine 98%, ee 94%, ChiPros®
(R)-(-)-α-Cyclohexanemethylamine
Purity: 98%
CAS No.: 5913-13-3
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 51°(123°F)
Refractive Index: 1.4620
Odor: Amine-like
pH: 11
Boiling Point: 177-178°
Beilstein Reference: 2935070
Optical Rotation: -4° (neat)
UN No.: 2733
MDL No.: MFCD00043338
Formula:
C₈H₁₇N MW: 127.23 g/mol Boiling Pt: 177…178 °C Density: 0.866 Flash Pt: 51 °C(123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043338 CAS Number: 5913-13-3 EINECS: 227-634-2 UN: 2733 ADR: "3",III |
Specification Test Results
Optical rotation | -4° (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...