(S)-(-)-3-Hydroxy-3-phenylpropionic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-3-Hydroxy-3-phenylpropanoic acid

L19041-03 L19041-MD
CAAAAL19041-03EA 316 CAD
CAAAAL19041-03 CAAAAL19041-MD
(S)-(-)-3-Hydroxy-3-phenylpropionic acid 98+%
(S)-(-)-3-Hydroxy-3-phenylpropionic acid

Purity: 98+% CAS No.: 36567-72-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 117-119° Beilstein Reference: 3197773

Formula: C₉H₁₀O₃
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00145262
CAS Number: 36567-72-3

Order Now

Specification Test Results

Optical rotation -22° (c=4 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR