(R)-(+)-3-Hydroxy-3-phenylpropionic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19040-03
L19040-MD
CAAAAL19040-03EA
322.32
CAD
CAAAAL19040-03
CAAAAL19040-MD
(R)-(+)-3-Hydroxy-3-phenylpropionic acid 98+%
(R)-(+)-3-Hydroxy-3-phenylpropionic acid
Purity: 98+%
CAS No.: 2768-42-5
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 118-121°
Beilstein Reference: 3197772
Formula:
C₉H₁₀O₃ Melting Pt: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145219 CAS Number: 2768-42-5 |
Specification Test Results
Optical rotation | +22° (c=4 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...