Ambersep® 900 (OH), Ambersep® ion exchange resin

Supplier: Thermo Scientific Chemicals

L19035-0B L19035-22 L19035-36
CAAAAL19035-0BEA 189.6 CAD
CAAAAL19035-0B CAAAAL19035-22 CAAAAL19035-36
Ambersep® 900 (OH), Ambersep® ion exchange resin
Ambersep® 900 (OH)

CAS No.: 9017-79-2 Odor: Odorless MDL No.: MFCD00802695

Storage Temperature: Ambient
MDL Number: MFCD00802695
CAS Number: 9017-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR