D-Epoxone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,2:4,5-Di-O-Isopropylidene-Beta-D-Erythro-2,3-Hexodiulo-2,6-Pyranose
L18989-03
L18989-06
CAAAAL18989-03EA
94.17
CAD
CAAAAL18989-03
CAAAAL18989-06
D-Epoxone 98%
D-Epoxone
Purity: 98%
CAS No.: 18422-53-2
Molecular Formula: C12H18O6
Formula Weight: 258.27
Melting Point: 101-103°
Beilstein Reference: 1319073
MDL No.: MFCD00063383
Formula:
C₁₂H₁₈O₆ MW: 258.27 g/mol Melting Pt: 101…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063383 CAS Number: 18422-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...