4-Isopropoxybenzenesulfonyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Isopropoxyphenylsulphonyl Chloride, 4-Isopropoxyphenylsulfonyl Chloride
L18984-03
L18984-06
CAAAAL18984-03EA
54.35
CAD
CAAAAL18984-03
CAAAAL18984-06
4-Isopropoxybenzenesulfonyl chloride 96%
4-Isopropoxybenzenesulfonyl chloride
Purity: 96%
CAS No.: 98995-40-5
Molecular Formula: C9H11ClO3S
Formula Weight: 234.70
Physical Form: Crystalline solid
Sensitivity: Moisture Sensitive
Appearance: White to pale cream
Melting Point: 30-37°
Boiling Point: 112-113°/0.01mm
Beilstein Reference: 3269058
UN No.: 3261
MDL No.: MFCD03093083
Formula:
C₉H₁₁ClO₃S MW: 234.69 g/mol Boiling Pt: 112…113 °C/0.01 mm Melting Pt: 30…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093083 CAS Number: 98995-40-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...