5-Methylnicotinamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18970-03
L18970-06
CAAAAL18970-03EA
110.44
CAD
CAAAAL18970-03
CAAAAL18970-06
5-Methylnicotinamide 97%
5-Methylnicotinamide
Purity: 97%
CAS No.: 70-57-5
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 166-168°
Beilstein Reference: 4962976
Formula:
C₇H₈N₂O MW: 136.15 g/mol Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664041 CAS Number: 70-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...