4-Hydroxydiphenylamine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-(Phenylamino)phenol
L18945-09
L18945-18
CAAAAL18945-09EA
70.47
CAD
CAAAAL18945-09
CAAAAL18945-18
4-Hydroxydiphenylamine 98%
4-Hydroxydiphenylamine
Purity: 98%
CAS No.: 122-37-2
Molecular Formula: C12H11NO
Formula Weight: 185.23
Flash Point: 209°(408°F)
Odor: Odorless
Boiling Point: 215-218°/12mm
Beilstein Reference: 511942
MDL No.: MFCD00020142
Formula:
C₁₂H₁₁NO MW: 185.23 g/mol Boiling Pt: 215…218 °C/12 mm Melting Pt: 66…71 °C Flash Pt: 209 °C(408 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020142 CAS Number: 122-37-2 EINECS: 204-538-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...