(R)-Methyl-2-amino-2-phenylacetate hydrochloride 96%

Supplier: Thermo Scientific Chemicals

Synonyms: D-(-)-2-Phenylglycine methyll ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, Methyl (R)-(-)-2-Amino-2-phenylacetate hydrochloride, H-D-Phg-OMe·HCl, H-D-Phg-OMe.HCl, (R)-(-)-2-Amino-2-phenylacetic acid methyll ester hydrochloride

L18735-14 L18735-22
CAAAAL18735-14EA 67.62 CAD
CAAAAL18735-14 CAAAAL18735-22
(R)-Methyl-2-amino-2-phenylacetate hydrochloride 96%
(R)-Methyl-2-amino-2-phenylacetate hydrochloride

Purity: 96% CAS No.: 19883-41-1 Molecular Formula: C9H11NO2.HCl Formula Weight: 201.65 Melting Point: ca 190° dec. Beilstein Reference: 3915654 Optical Rotation: -118° (c=1 in water) MDL No.: MFCD00137487

Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
Melting Pt: ∼ 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00137487
CAS Number: 19883-41-1
EINECS: 243-399-9

Order Now

Specification Test Results

Optical rotation -118° (c=1 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR