2-Allyl-4,6-dibenzoylresorcinol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18722-06
L18722-14
CAAAAL18722-06EA
70.15
CAD
CAAAAL18722-06
CAAAAL18722-14
2-Allyl-4,6-dibenzoylresorcinol 98%
2-Allyl-4,6-dibenzoylresorcinol
Purity: 98%
CAS No.: 102593-74-8
Molecular Formula: C23H18O4
Formula Weight: 358.40
Melting Point: 158-163°
Beilstein Reference: 2168154
MDL No.: MFCD02094038
Formula:
C₂₃H₁₈O₄ Melting Pt: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD02094038 CAS Number: 102593-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...