Methyl-3-isocyanatobenzoate 97%

Supplier: Thermo Scientific Chemicals
Danger

L18709-03 L18709-06
CAAAAL18709-03EA 102.54 CAD
CAAAAL18709-03 CAAAAL18709-06
Methyl-3-isocyanatobenzoate 97%
Methyl 3-isocyanatobenzoate

Purity: 97% CAS No.: 41221-47-0 Molecular Formula: C9H7NO3 Formula Weight: 177.16 Flash Point: 110°(230°F) Melting Point: 35-38° UN No.: 2811 MDL No.: MFCD00037077

Formula: C₉H₇NO₃
Melting Pt: 35…38 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037077
CAS Number: 41221-47-0
UN: 3335
ADR: "9",

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR